Anti-L1CAM Mouse Monoclonal Antibody [clone: 2A6]

Supplier: OriGene
ORIGCF500728EA 749 EUR
ORIGCF500728
Anti-L1CAM Mouse Monoclonal Antibody [clone: 2A6]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR