Anti-DHPS Mouse Monoclonal Antibody [clone: 2C9]

Supplier: OriGene
ORIGCF501393EA 749 EUR
ORIGCF501393
Anti-DHPS Mouse Monoclonal Antibody [clone: 2C9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DHPS
Clonality: Monoclonal
Clone: 2C9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR