Anti-STK3 Mouse Monoclonal Antibody [clone: 2D4]

Supplier: OriGene
ORIGCF500409EA 749 EUR
ORIGCF500409
Anti-STK3 Mouse Monoclonal Antibody [clone: 2D4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: STK3
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR