Anti-SILV Mouse Monoclonal Antibody [clone: 7B2]

Supplier: OriGene
ORIGCF500423EA 749 EUR
ORIGCF500423
Anti-SILV Mouse Monoclonal Antibody [clone: 7B2]
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: SILV
Clonality: Monoclonal
Clone: 7B2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR