Anti-SSB Mouse Monoclonal Antibody [clone: 6G7]

Supplier: OriGene
ORIGCF500427EA 749 EUR
ORIGCF500427
Anti-SSB Mouse Monoclonal Antibody [clone: 6G7]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SSB
Clonality: Monoclonal
Clone: 6G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR