Anti-CHCHD5 Mouse Monoclonal Antibody [clone: 3H3]

Supplier: OriGene
ORIGCF502399EA 749 EUR
ORIGCF502399
Anti-CHCHD5 Mouse Monoclonal Antibody [clone: 3H3]
Antibodies
Recommended Dilutions: Western Blot: 1:200; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CHCHD5
Clonality: Monoclonal
Clone: 3H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR