Anti-SORD Mouse Monoclonal Antibody [clone: 17E7]

Supplier: OriGene
ORIGCF500748EA 749 EUR
ORIGCF500748
Anti-SORD Mouse Monoclonal Antibody [clone: 17E7]
Antibodies
Recommended Dilutions: Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: SORD
Clonality: Monoclonal
Clone: 17E7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR