Anti-RPA2 Mouse Monoclonal Antibody [clone: 9A1]

Supplier: OriGene
ORIGCF500763EA 749 EUR
ORIGCF500763
Anti-RPA2 Mouse Monoclonal Antibody [clone: 9A1]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: RPA2
Clonality: Monoclonal
Clone: 9A1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR