Anti-PKLR Mouse Monoclonal Antibody [clone: 1G9]
Supplier: OriGene
ORIGCF501418EA 749 EUR
ORIGCF501418
Anti-PKLR Mouse Monoclonal Antibody [clone: 1G9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: PKLR
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human