Anti-NRBP1 Mouse Monoclonal Antibody [clone: 2A5]

Supplier: OriGene
ORIGCF500449EA 749 EUR
ORIGCF500449
Anti-NRBP1 Mouse Monoclonal Antibody [clone: 2A5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NRBP1
Clonality: Monoclonal
Clone: 2A5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR