Anti-MAPK1 Mouse Monoclonal Antibody [clone: 7C9]

Supplier: OriGene
ORIGCF500473EA 749 EUR
ORIGCF500473
Anti-MAPK1 Mouse Monoclonal Antibody [clone: 7C9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 7C9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR