Anti-UBL4A Mouse Monoclonal Antibody [clone: 3C12]

Supplier: OriGene
ORIGCF502448EA 749 EUR
ORIGCF502448
Anti-UBL4A Mouse Monoclonal Antibody [clone: 3C12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: UBL4A
Clonality: Monoclonal
Clone: 3C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR