Anti-GBE1 Mouse Monoclonal Antibody [clone: 3B4]

Supplier: OriGene
ORIGCF500801EA 749 EUR
ORIGCF500801
Anti-GBE1 Mouse Monoclonal Antibody [clone: 3B4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: GBE1
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR