Anti-AKT2 Mouse Monoclonal Antibody [clone: 8G8]

Supplier: OriGene
ORIGCF500808EA 749 EUR
ORIGCF500808
Anti-AKT2 Mouse Monoclonal Antibody [clone: 8G8]
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 8G8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR