Anti-ID3 Mouse Monoclonal Antibody [clone: 7B6]

Supplier: OriGene
ORIGCF500811EA 749 EUR
ORIGCF500811
Anti-ID3 Mouse Monoclonal Antibody [clone: 7B6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: ID3
Clonality: Monoclonal
Clone: 7B6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR