Anti-PRL Mouse Monoclonal Antibody [clone: 2C3]

Supplier: OriGene
ORIGCF500818EA 770 EUR
ORIGCF500818
Anti-PRL Mouse Monoclonal Antibody [clone: 2C3]
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PRL
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR