Anti-AFP Mouse Monoclonal Antibody [clone: 2E2]

Supplier: OriGene
ORIGCF501788EA 749 EUR
ORIGCF501788
Anti-AFP Mouse Monoclonal Antibody [clone: 2E2]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: 2E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR