Anti-FERMT2 Mouse Monoclonal Antibody [clone: 25A4]

Supplier: OriGene
ORIGCF500507EA 749 EUR
ORIGCF500507
Anti-FERMT2 Mouse Monoclonal Antibody [clone: 25A4]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: FERMT2
Clonality: Monoclonal
Clone: 25A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR