Anti-ERCC4 Mouse Monoclonal Antibody [clone: 6E6]

Supplier: OriGene
ORIGCF503294EA 749 EUR
ORIGCF503294
Anti-ERCC4 Mouse Monoclonal Antibody [clone: 6E6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC4
Clonality: Monoclonal
Clone: 6E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR