Anti-PDE6G Mouse Monoclonal Antibody [clone: 1H7]

Supplier: OriGene
ORIGCF503705EA 749 EUR
ORIGCF503705
Anti-PDE6G Mouse Monoclonal Antibody [clone: 1H7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PDE6G
Clonality: Monoclonal
Clone: 1H7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR