Anti-PSMB7 Mouse Monoclonal Antibody [clone: 1B2]

Supplier: OriGene
ORIGCF504456EA 770 EUR
ORIGCF504456
Anti-PSMB7 Mouse Monoclonal Antibody [clone: 1B2]
Antibodies
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: PSMB7
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR