Anti-CNOT4 Mouse Monoclonal Antibody [clone: 3A3]

Supplier: OriGene
ORIGCF504472EA 749 EUR
ORIGCF504472
Anti-CNOT4 Mouse Monoclonal Antibody [clone: 3A3]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CNOT4
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR