Anti-DPH2 Mouse Monoclonal Antibody [clone: 4D9]
Supplier: OriGene
ORIGCF504853EA 770 EUR
ORIGCF504853
Anti-DPH2 Mouse Monoclonal Antibody [clone: 4D9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: DPH2
Clonality: Monoclonal
Clone: 4D9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human