Anti-ACBD3 Mouse Monoclonal Antibody [clone: 2B12]

Supplier: OriGene
ORIGCF504857EA 749 EUR
ORIGCF504857
Anti-ACBD3 Mouse Monoclonal Antibody [clone: 2B12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ACBD3
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR