Anti-PADI4 Mouse Monoclonal Antibody [clone: 5C10]

Supplier: OriGene
ORIGCF504896EA 749 EUR
ORIGCF504896
Anti-PADI4 Mouse Monoclonal Antibody [clone: 5C10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PADI4
Clonality: Monoclonal
Clone: 5C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR