Anti-FTCD Mouse Monoclonal Antibody [clone: 4B6]

Supplier: OriGene
ORIGCF504947EA 770 EUR
ORIGCF504947
Anti-FTCD Mouse Monoclonal Antibody [clone: 4B6]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: FTCD
Clonality: Monoclonal
Clone: 4B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR