Imprimer

Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 3B4]

Fournisseur: OriGene
ORIGCF505728EA 749 EUR
ORIGCF505728
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 3B4]
Anticorps
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR