Anti-LPL Mouse Monoclonal Antibody [clone: 6B4]
Supplier: OriGene
ORIGCF503784EA 749 EUR
ORIGCF503784
Anti-LPL Mouse Monoclonal Antibody [clone: 6B4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: LPL
Clonality: Monoclonal
Clone: 6B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human