Anti-PDIA3 Mouse Monoclonal Antibody [clone: 2D2]

Supplier: OriGene
ORIGCF504993EA 749 EUR
ORIGCF504993
Anti-PDIA3 Mouse Monoclonal Antibody [clone: 2D2]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: PDIA3
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR