Anti-MMACHC Mouse Monoclonal Antibody [clone: 4E12]

Supplier: OriGene
ORIGCF504998EA 770 EUR
ORIGCF504998
Anti-MMACHC Mouse Monoclonal Antibody [clone: 4E12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MMACHC
Clonality: Monoclonal
Clone: 4E12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR