Anti-PDIA3 Mouse Monoclonal Antibody [clone: 3H5]

Supplier: OriGene
ORIGCF505010EA 749 EUR
ORIGCF505010
Anti-PDIA3 Mouse Monoclonal Antibody [clone: 3H5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PDIA3
Clonality: Monoclonal
Clone: 3H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR