Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 2B2]

Supplier: OriGene
ORIGCF505745EA 749 EUR
ORIGCF505745
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 2B2]
Antibodies
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 2B2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR