Imprimer

Anti-NUDT18 Mouse Monoclonal Antibody [clone: 5F9]

Fournisseur: OriGene
ORIGCF503865EA 770 EUR
ORIGCF503865
Anti-NUDT18 Mouse Monoclonal Antibody [clone: 5F9]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NUDT18
Clonality: Monoclonal
Clone: 5F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR