Anti-RFC2 Mouse Monoclonal Antibody [clone: 3F6]

Supplier: OriGene
ORIGCF505037EA 770 EUR
ORIGCF505037
Anti-RFC2 Mouse Monoclonal Antibody [clone: 3F6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: RFC2
Clonality: Monoclonal
Clone: 3F6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR