Anti-CTAG1B Mouse Monoclonal Antibody [clone: 5B2]

Supplier: OriGene
ORIGCF505949EA 749 EUR
ORIGCF505949
Anti-CTAG1B Mouse Monoclonal Antibody [clone: 5B2]
Antibodies
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: CTAG1B
Clonality: Monoclonal
Clone: 5B2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR