Anti-FXYD3 Mouse Monoclonal Antibody [clone: 1E8]

Supplier: OriGene
ORIGCF504717EA 749 EUR
ORIGCF504717
Anti-FXYD3 Mouse Monoclonal Antibody [clone: 1E8]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: FXYD3
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR