Anti-SCFD1 Mouse Monoclonal Antibody [clone: 5G4]

Supplier: OriGene
ORIGCF504721EA 749 EUR
ORIGCF504721
Anti-SCFD1 Mouse Monoclonal Antibody [clone: 5G4]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: SCFD1
Clonality: Monoclonal
Clone: 5G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR