Anti-MPI Mouse Monoclonal Antibody [clone: 1C7]

Supplier: OriGene
ORIGCF504749EA 749 EUR
ORIGCF504749
Anti-MPI Mouse Monoclonal Antibody [clone: 1C7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MPI
Clonality: Monoclonal
Clone: 1C7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR