Anti-BBOX1 Mouse Monoclonal Antibody [clone: 2F4]

Supplier: OriGene
ORIGCF505789EA 749 EUR
ORIGCF505789
Anti-BBOX1 Mouse Monoclonal Antibody [clone: 2F4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: BBOX1
Clonality: Monoclonal
Clone: 2F4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR