Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 2A3]

Supplier: OriGene
ORIGCF505794EA 749 EUR
ORIGCF505794
Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 2A3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PIK3R5
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR