Anti-MAPK4 Mouse Monoclonal Antibody [clone: 2D7]

Supplier: OriGene
ORIGCF505808EA 749 EUR
ORIGCF505808
Anti-MAPK4 Mouse Monoclonal Antibody [clone: 2D7]
Antibodies
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAPK4
Clonality: Monoclonal
Clone: 2D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR