Anti-USP7 Mouse Monoclonal Antibody [clone: 1F12]

Supplier: OriGene
ORIGCF503987EA 749 EUR
ORIGCF503987
Anti-USP7 Mouse Monoclonal Antibody [clone: 1F12]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: USP7
Clonality: Monoclonal
Clone: 1F12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR