Anti-SLA2 Mouse Monoclonal Antibody [clone: 3D4]

Supplier: OriGene
ORIGCF505152EA 749 EUR
ORIGCF505152
Anti-SLA2 Mouse Monoclonal Antibody [clone: 3D4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SLA2
Clonality: Monoclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR