Anti-UBE2S Mouse Monoclonal Antibody [clone: 1B5]

Supplier: OriGene
ORIGCF505168EA 749 EUR
ORIGCF505168
Anti-UBE2S Mouse Monoclonal Antibody [clone: 1B5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: UBE2S
Clonality: Monoclonal
Clone: 1B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR