Imprimer

Anti-PROSC Mouse Monoclonal Antibody [clone: 2H1]

Fournisseur: OriGene
ORIGCF505194EA 749 EUR
ORIGCF505194
Anti-PROSC Mouse Monoclonal Antibody [clone: 2H1]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PROSC
Clonality: Monoclonal
Clone: 2H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR