Anti-GZMB Mouse Monoclonal Antibody [clone: 4E6]

Supplier: OriGene
ORIGCF505930EA 749 EUR
ORIGCF505930
Anti-GZMB Mouse Monoclonal Antibody [clone: 4E6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: GZMB
Clonality: Monoclonal
Clone: 4E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR