Anti-CDKN2B Mouse Monoclonal Antibody [clone: 3B6]

Supplier: OriGene
ORIGCF505206EA 749 EUR
ORIGCF505206
Anti-CDKN2B Mouse Monoclonal Antibody [clone: 3B6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CDKN2B
Clonality: Monoclonal
Clone: 3B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR