Anti-HDHD2 Mouse Monoclonal Antibody [clone: 1A4]

Supplier: OriGene
ORIGCF502885EA 749 EUR
ORIGCF502885
Anti-HDHD2 Mouse Monoclonal Antibody [clone: 1A4]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: HDHD2
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR