Anti-ALDOB Mouse Monoclonal Antibody [clone: 3C6]

Supplier: OriGene
ORIGCF502908EA 749 EUR
ORIGCF502908
Anti-ALDOB Mouse Monoclonal Antibody [clone: 3C6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ALDOB
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR