Anti-MAPRE2 Mouse Monoclonal Antibody [clone: 4G1]

Supplier: OriGene
ORIGCF502918EA 749 EUR
ORIGCF502918
Anti-MAPRE2 Mouse Monoclonal Antibody [clone: 4G1]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MAPRE2
Clonality: Monoclonal
Clone: 4G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR